Neodymium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide

11248-18 11248-30 11248-A1
CAAA11248-18EA 165.9 CAD
CAAA11248-18 CAAA11248-30 CAAA11248-A1
Neodymium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Neodymium (III) oxide

Powder. Purity based on rare earth contaminates.


Purity: 99.9% CAS No.: 1313-97-9 Molecular Formula: Nd2O3 Formula Weight: 336.48 Physical Form: Powder Density: 7.24 Melting Point: 2272° Merck Reference: 14,6450 MDL No.: MFCD00011134

Formula: Nd₂O₃
MW: 336.48 g/mol
Boiling Pt: 3760 °C (1013 hPa)
Melting Pt: 2272 °C
Density: 7.24
Storage Temperature: Ambient
MDL Number: MFCD00011134
CAS Number: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,6450

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR