Methyl stearate ≥97.0%

Supplier: TCI America

Synonyms: Stearic acid methyl ester, Methyl octadecanoate

S0080-500G S0080-25G
TCS0080-500GEA 97.61 CAD
TCS0080-500G TCS0080-025G
Methyl stearate ≥97.0%
Methyl stearate

CAS Number: 112-61-8
MDL Number: MFCD00009005
Molecular Formula: C19H38O2
Molecular Weight: 298.51
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 182
Melting point (°C): 40
Flash Point (°C): 153


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Stearic Acid Methyl Ester

Formula: CH₃(CH₂)₁₆CO₂CH₃
MW: 298.51 g/mol
Boiling Pt: 182 °C
Melting Pt: 40 °C
Density: 1.007 g/cm³ (20 °C)
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00009005
CAS Number: 112-61-8
EINECS: 203-990-4
Merck Index: 12,08959
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c266bd0d80747c6ccfcf9bb1e529834c1b8c776&vid=3c266bd0d80747c6ccfcf9bb1e529834c1b8c776&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR