VWR® Sticky Mat
Supplier: VWR International
77461-300
77461-300EA 81.7 CAD
77461-300
VWR® Sticky Mat
Shakers Shaking Incubators
Easily secure and remove samples without clamps.
  • Recommemded maximum speed not to exceed 250 rpm
  • Application temperature not to exceed 65 °C
  • Clean with water and air dry to restore adhesive quality