NEXTFLEX™ Human Core Exome Kits
Supplier: Revvity Health Sciences
NEXTFLEX™
NOVA-6011-EVAL NOVA-6011-48 NOVA-6011-EVAL NOVA-6011-96 NOVA-6011-48 NOVA-6011-96
CA77448-222EA 5607.93 CAD
CA77448-222 77448-224 77448-222 CA77448-226 CA77448-224 77448-226
NEXTFLEX™ Human Core Exome Kits
Nucleic Acid Reagents Next Generation Sequencing Reagents
NEXTFLEX® Human core exome V2 (CES) panel is designed to overcome the limitations of analyzing genetic variants of interest with respect to conventional whole-exome sequencing.
  • Complete whole exome coverage
  • Exceptional target capture performance
  • Rapid same-day workflow
  • Automated on Sciclone® G3 NGSx workstation
  • Bead-based normalization
The NEXTFLEX® human CES panel includes significant genes used in other panels and additional genes, therefore allowing customers to obtain comprehensive information of mutations in genes of interest in a cost-effective manner. The 7513 genes covered in this panel can be reviewed in the NEXTFLEX CES panel gene list.
Certifications: Manufactured in ISO 9001 Facility.
Caution: For research use only. Not for use in diagnostic procedures.