NEXTFLEX™ Rapid DNA-seq 2.0 Kits
Supplier: REVVITY HEALTH SCIENCES CANADA CA
The NEXTFLEX® Rapid DNA-seq 2.0 kit is designed to prepare multiplexed libraries for single or paired-end sequencing using Illumina® platforms from fragmented DNA.
- Complete library prep solution, including size selection beads
- Up to 1536 UDI barcodes are available separately
- Convenient and cost-effective package in an 8, 48, or 96 reaction format
- Simplified workflow
- Robust genome coverage
- Reliable performance with GC-bias and sequencing bias mitigation
- Low input requirement for genomic DNA down to 1 ng
- Functionally tested with Illumina® sequencing platform
- Automation compatible with liquid handlers, including Revvity's Sciclone® G3 NGSx workstations, Zephyr® G3 NGS workstation, and BioQule NGS system
The NEXTFLEX Rapid DNA-seq kit 2.0 produces library preps with a larger number of unique sequencing reads, allowing for robust genome coverage in challenging GC-rich sequences that can introduce bias.
Certifications: Manufactured in ISO 9001 Facility.
Caution: For research use only. Not for use in diagnostic procedures.