Polycarbin® LTS® Compatible Refill Towers
Supplier: POLYCARBIN, INC.
PC01-04-06R PC01-05-06R PC01-06-06R
77461-176CS 341.99 CAD
77461-176 77461-178 77461-180 77461-182
Polycarbin® LTS® Compatible Refill Towers
Pipette Tips
Polycarbin's LTS® compatible refill towers deliver the precision and accuracy you expect, with a fraction of the carbon footprint.
  • Wafer and rack made from 100% circular economy, post-consumer recycled content
  • Smaller carbon footprint compared to other refill and rack systems
  • Easy to transfer refills into polycarbin racks
  • Certified DNase, RNase and pyrogen free
  • Non-sterile (autoclavable)
Polycarbin's LTS® compatible refill system is designed to deliver an exceptional product that is less carbon and plastic-intensive. Refill tower housing is manufactured using post-consumer recycled content, and are shrink wrapped and packaged in an easy to recycle cardboard sleeve. Pipette tips do not contain recycled content and are made from virgin PP resin.
Certifications: Certified sterile and DNase, RNase and pyrogen free. Manufactured in an ISO 9001 compliant facility.
Ordering information: Note that Polycarbin's LTS® compatible refill towers do not come with racks (bases). These can be ordered separately by the sleeve. Please see part numbers 76635-484, 76635-488 and 76635-492.