VWR® ESD Cleanroom Polyester Wipes
Supplier: VWR International
77395-162 77395-164
77395-162BG 38.68 CAD
77395-162 77395-164
VWR® ESD Cleanroom Polyester Wipes
Wipes Disposable Wipes and Towels Wipes for General Applications
VWR® ESD Cleanroom Wipers are specially designed for use in environments where electrostatic discharge (ESD) protection is necessary, such as in semiconductor manufacturing, electronics assembly, and cleanroom environments. These wipers are made from materials that are static dissipative, preventing the buildup of static electricity that can damage sensitive electronic components. ESD cleanroom wipers are essential tools for maintaining cleanliness and ESD protection in sensitive environments.
  • For use in most critical cleanroom applications
  • 100% continuous filament polyester wiper
  • Edges sealed with patented laser-cutting technology
  • Exceptional absorbency and cleaning efficiency
  • Laundered with 18MΩ DI water
  • Low fiber and particulate release
The VWR® ESD Cleanroom wiper is a 100% continuous filament polyester wiper, featuring a double-knit construction. The wipe edges are sealed with laser cutting technology, resulting in very low fiber and particle release for the most critical cleanroom applications.