Sabouraud Dextrose Agar Plates
Supplier: TEKNOVA CA
S2050 S2040
CA100217-298PK 972.24 CAD
CA100217-298 CA100220-142
Sabouraud Dextrose Agar Plates
Media for Microbiology Prepared Media for Microbiology
Sabouraud Dextrose is used for the qualitative cultivation of dermatophytes, yeasts, molds and aciduric microorganisms.
  • 20 plates
  • Sterile
Sabouraud dextrose agar recipe (per Liter): 1.0% Tryptone (10.0 g/L), 4.0% Dextrose (40.0 g/L), 1.5% Agar (15.0 g/L), Final pH: 5.6 ±0.2 (at 25 °C).
Ambient shipping conditions. Store at room temperature.