Battery Hydrometer and Syphon Set
Supplier: Heathrow Scientific
6605-5 6605-5H 6605-OSY
76546-440EA 147.94 CAD
76546-440 76546-442 76546-444
Battery Hydrometer and Syphon Set
Hydrometers
Our battery acid hydrometer set is used in many industrial settings including fleet maintenance, automotive services, solar-powered system maintenance, and general industrial applications.
  • The rubber parts are made of nitrile rubber, which is chemically inert and durable
  • Calibrated for use at 77 °F
A reinforced box provides storage and is compartmented to eliminate breakage.
Delivery information: The set includes a specific gravity hydrometer and glass syphon tube with a rubber bulb, nozzle, and clamp.