Armored Encapsulated Glass Thermometers
Supplier: Heathrow Scientific
80701E-A 80702E-A 80703E-A 80704E-A 80706E-A 80707E-A 80708E-A 80901E-A 80902E-A 80903E-A 80904E-A 80906E-A 80907E-A 80908E-A
76546-610EA 122.72 CAD
76546-610 76546-616 76546-622 76546-626 76546-632 76546-638 76546-644 76546-650 76546-656 76546-662 76546-666 76546-674 76546-680 76546-686
Armored Encapsulated Glass Thermometers
Thermometers Analog Thermometers
Our precision, serialized thermometers are encapsulated in FEP. The armor is constructed of nickel-plated brass.
  • Includes FEP encapsulation and nickel-plated brass armor
  • Feature safety blue fill against a white background for easy reading
  • Wide variety of temperature ranges available
These are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications. In the event of breakage the FEP coating contains the broken glass and liquid fill, preventing injury or contamination. The armor protects the thermometer from rough handling and shock during use.