Serialized Armored Glass Thermometers
Supplier: Heathrow Scientific
80701-A 80702-A 80703-A 80706-A 80707-A 80708-A 80901-A 80902-A 80903-A 80905-A 80906-A 80907-A 80908-A 80910-A
76546-606EA 71.48 CAD
76546-606 76546-612 76546-618 76546-628 76546-634 76546-640 76546-646 76546-652 76546-658 76546-668 76546-670 76546-676 76546-682 76546-688
Serialized Armored Glass Thermometers
Thermometers Analog Thermometers
Our precision, serialized thermometers include an armor that is a 2 part mickel plated brass.
  • Nickel plated brass armor
  • Feature safety blue fill against a white background for easy reading
  • Wide variety of temperature ranges available
These are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications requiring serialization.