2-Formyl-6-picoline ≥98.0%

Supplier: TCI America

Synonyms: 6-Methylpyridine-2-carboxaldehyde, 6-Methyl-2-pyridinecarboxaldehyde

M1157-5G M1157-25G
TCM1157-005GEA 101.12 CAD
TCM1157-005G TCM1157-025G
2-Formyl-6-picoline ≥98.0%
2-Formyl-6-picoline

CAS Number: 1122-72-1
MDL Number: MFCD00006291
Molecular Formula: C7H7NO
Molecular Weight: 121.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 68
Freezing point (°C): 30
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Formyl-6-methylpyridine, 6-Methylpicolinaldehyde

Formula: C₇H₇NO
Flash Pt: 68 °C
MDL Number: MFCD00006291
CAS Number: 1122-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3c04068f17547517792ed471f90e2234b718c07&vid=c3c04068f17547517792ed471f90e2234b718c07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR