Ethyl hydrogen maleate ≥95.0%

Supplier: TCI America

Synonyms: Monoethyl Maleate, Maleic Acid Monoethyl Ester

M0614-25ML M0614-500ML M0614-100ML
TCM0614-025MLEA 139.04 CAD
TCM0614-025ML TCM0614-500ML TCM0614-100ML
Ethyl hydrogen maleate ≥95.0%
Ethyl hydrogen maleate

CAS Number: 3990-03-2
MDL Number: MFCD00063175
Molecular Formula: C6H8O4
Molecular Weight: 144.13
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 1.18
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Maleic Acid Monoethyl Ester, Monoethyl Maleate

Formula: C₆H₈O₄
MW: 144.13 g/mol
Density: 1.18 (20 °C)
MDL Number: MFCD00063175
CAS Number: 3990-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=023d9524045626e8af5ee8c1702b3a48244c642e&vid=023d9524045626e8af5ee8c1702b3a48244c642e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR