Round-Bottom Three-Neck Flasks, Vertical Necks, Standard Walls, Ace Glass Incorporated
Supplier: Ace Glass
6944-03 6944-04 6944-05 6944-06 6944-10 6944-148 6944-157 6944-16 6944-167 6944-177 6944-178 6944-18 6944-188 6944-20 6944-22 6944-24 6944-25 6944-26 6944-28 6944-30 6944-31 6944-38 6944-40 6944-42 6944-44 6944-46 6944-47 6944-48 6944-49 6944-50 6944-54 6944-56 6944-57 6944-58 6944-59 6944-60 6944-62 6944-64 6944-66 6944-67 6944-68 6944-69 6944-70 6944-71 6944-72 6944-73 6944-74 6944-75 6944-76 6944-78 6944-79 6944-80 6944-81 6944-82 6944-83 6944-84 6944-86 6944-88 6944-91 6944-107 6944-109 6944-111 6944-113 6944-115 6944-117 6944-119 6944-120 6944-127
89056-580EA 360.94 CAD
89056-580 89056-582 89056-584 89056-586 89056-588 89056-590 89056-592 89056-594 89056-596 89056-598 89056-600 89056-602 89056-604 89056-606 89056-622 89056-646 89056-664 89056-678 89056-692 89056-698 89056-700 89056-702 89056-704 89056-706 89056-708 89056-710 89056-712 89056-714 89056-716 89056-718 89056-720 89056-722 89056-724 89056-726 89056-728 89056-730 89056-732 89056-734 89056-736 89056-738 89056-740 89056-742 89056-744 89056-746 89056-748 89056-750 89056-752 89056-754 89056-756 89056-758 89056-760 89056-762 89056-764 89056-766 89056-768 89056-770 89056-772 89056-774 89056-776 89083-886 89083-890 89083-892 89083-898 89083-900 89083-908 89083-910 89083-912 89083-918
Round-Bottom Three-Neck Flasks, Vertical Necks, Standard Walls, Ace Glass Incorporated
Flasks Round Bottom Flasks
Round-bottom flasks with three vertical necks and [ST] outer joints
The outer joints are tooled with reinforcement rings (full-sized joints only) for added strength and stability. The flare-out directly below the center joint facilitates cleaning of the flask and enables stirring blades to be easily removed. Fabricated from borosilicate glass.