Methacrylic acid-tert-butyl ester ≥98% stabilised

Supplier: TCI America
Warning

Synonyms: Methacrylic acid tert-Butyl ester monomer, tert-Butyl Methacrylate

M0326-500ML M0326-25ML
TCM0326-500MLEA 270.59 CAD
TCM0326-500ML TCM0326-025ML
Methacrylic acid-tert-butyl ester ≥98% stabilised
Methacrylic acid-tert-butyl ester

(stabilized with MEHQ)
CAS Number: 585-07-9
MDL Number: MFCD00048245
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 30
Specific Gravity (20/20): 0.88


Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents
Other Product Names / Synonyms: Methacrylic Acid tert-Butyl Ester Monomer

Formula: C₈H₁₄O₂
MW: 142.1956 g/mol
Boiling Pt: 136 °C
Density: 0.88 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00048245
CAS Number: 585-07-9
UN: 3272
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0999f8589f3633e27a7f6dda8463f7bffde3fa6&vid=a0999f8589f3633e27a7f6dda8463f7bffde3fa6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR