N-Methylacetamide ≥99.0%
Supplier: TCI America
M0133-25G
M0133-250G
TCM0133-025GEA
30.56
CAD
TCM0133-025G
TCM0133-250G
N-Methylacetamide ≥99.0%
N-Methylacetamide
CAS Number: 79-16-3
MDL Number: MFCD00008683
Molecular Formula: C3H7NO
Molecular Weight: 73.10
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 206
Flash Point (°C): 108
Freezing point (°C): 30
Other Product Names / Synonyms: N-Acetyl-N-methylamine
Formula:
C₃H₇NO Boiling Pt: 206 °C Flash Pt: 108 °C |
MDL Number:
MFCD00008683 CAS Number: 79-16-3 EINECS: 201-182-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...