2-Hydroxyethyl methacrylate ≥95.0% stabilised

Supplier: TCI America
Warning

Synonyms: HEMA, GMA, Methacrylic acid 2-hydroxyethyl ester, 1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate

M0085-25G M0085-500G
TCM0085-025GEA 34.76 CAD
TCM0085-025G TCM0085-500G
2-Hydroxyethyl methacrylate ≥95.0% stabilised
2-Hydroxyethyl methacrylate

(stabilized with MEHQ)
CAS Number: 868-77-9
MDL Number: MFCD00002863
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 107
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: Ethylene Glycol Methacrylate, Methacrylic Acid 2-Hydroxyethyl Ester

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 85 °C
Density: 1.07 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00002863
CAS Number: 868-77-9
EINECS: 212-782-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6ef0d6f114e8ee83a791040317f6a5024fc17db&vid=f6ef0d6f114e8ee83a791040317f6a5024fc17db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR