D-(+)-Malic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid

M0021-5G M0021-25G
TCM0021-005GEA 40.27 CAD
TCM0021-005G TCM0021-025G
D-(+)-Malic acid ≥98.0%
D(+)-Malic acid

CAS Number: 636-61-3
MDL Number: MFCD00004245
Molecular Formula: C4H6O5
Molecular Weight: 134.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 103
Specific rotation [a]20/D: 6.5 deg (C=10, Acetone)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks
Other Product Names / Synonyms: D-(+)-Apple Acid

Formula: C₄H₆O₅
MW: 134.09 g/mol
Boiling Pt: 322
Melting Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00004245
CAS Number: 636-61-3
EINECS: 211-262-2
Merck Index: 13,05307
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27f3df43216597a07a310ac9fce30582fefa6e2b&vid=27f3df43216597a07a310ac9fce30582fefa6e2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR