2-(N-Methylanilino)ethanol ≥97.0%

Supplier: TCI America

Synonyms: N-Methyl-N-2-ethanolaniline, 2-(Methyl(phenyl)amino)ethanol, N-(2-Hydroxyethyl)-N-methylaniline

H0359-25ML H0359-500ML
TCH0359-025MLEA 85.32 CAD
TCH0359-025ML TCH0359-500ML
2-(N-Methylanilino)ethanol ≥97.0%
2-(N-Methylanilino)ethanol

CAS Number: 93-90-3
MDL Number: MFCD00020572
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 266
Flash Point (°C): 110
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: N-(2-Hydroxyethyl)-N-methylaniline, N-Methyl-N-2-ethanolaniline

Formula: C₉H₁₃NO
Boiling Pt: 266 °C
Density: 1.07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00020572
CAS Number: 93-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b95861dbf28978959ab97d965d27b214b7c2c1&vid=77b95861dbf28978959ab97d965d27b214b7c2c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR