Round-Bottom Flasks with Threaded Side Port, Ace Glass Incorporated
Supplier: Ace Glass
6936-56 6936-230 6936-235 6936-237 6936-240 6936-242 6936-244 6936-246 6936-30 6936-35 6936-40 6936-44 9462-07 9462-09 9462-11 9462-13 9462-207 9462-209 9462-211
71001-548EA 270.13 CAD
71001-548 89056-548 89056-550 89056-552 89056-554 89056-556 89056-558 89056-560 89056-562 89056-564 89056-566 89056-568 89074-670 89074-672 89074-674 89074-676 89074-678 89074-680 89074-682
Round-Bottom Flasks with Threaded Side Port, Ace Glass Incorporated
Flasks Round Bottom Flasks
Round-bottom flasks with [ST] outer joint center neck and Ace-Thred side port
The outer joint is tooled with reinforcement rings (full-sized joints only) for added strength and stability. The internally threaded side port can accommodate thermometers or bleed tubes. Available in heavy or standard wall. Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks. Ace-Thred side port size (below) also refers to the outside diameter of tubing or thermometer that will fit this joint. Fabricated from borosilicate glass.
Ordering information: Supplied with nylon bushing and FETFE O-ring.