Methyl purple solution indicator

Supplier: Ricca Chemical

R5040000-500C R5040000-1C R5040000-120C
RC504016EA 86.32 CAD
RC504016 RC504032 RC50404
Methyl purple solution indicator
Methyl purple

Aqueous.

Formula: C₆₇H₆₂N₈Na₄O₁₄S₃
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0050 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00070843
CAS Number: 1340-02-9

Order Now

Specification Test Results

Appearance Clear, dark, medium green, odorless
Suitability for Use Purple (pH 4.8) - Green (pH 5.4), with Gray warning color


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR