Trifluoroacetic acid ≥99.9% for HPLC, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: TFA

M-Clarity™ quality level = MQ200

TX1276-0004 TX1276-4 TX1276-6 TX1276-7
CATX1276-0004EA 242 CAD
CATX1276-0004 CATX1276-4 CATX1276-6 CATX1276-7
Trifluoroacetic acid ≥99.9% for HPLC, Supelco®
Trifluoroacetic acid

Colorless to light yellow liquid.

Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.9% min
Chloride (Cl) 0.001% max
Color (ALPHA)5 max
Evaporation residue 0.005% max
Fluoride (F) 0.001% max
Form Clear liquid
Iron (Fe) 0.0001% max
Sulfate (SO4) 0.001% max
UV Absorbance (0.1%, 1 cm, H2O, 230 nm) 0.15AU max
UV Absorbance (0.1%, 1 cm, H2O, 254 nm) 0.01AU max
Water (H2O) 0.05% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR