Trifluoroacetic acid ≥99.5%, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
TX1275-1
TX1275-3
CATX1275-1EA
188
CAD
CATX1275-1
CATX1275-3
Trifluoroacetic acid ≥99.5%, Supelco®
Trifluoroacetic acid
Meets Reagent Specifications for testing USP/NF monographs
Formula:
C₂HF₃O₂ Boiling Pt: 71.8 °C (1013 hPa) Melting Pt: –15 °C Density: 1.490 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 EINECS: 200-929-3 UN: 2699 ADR: 8,I Merck Index: 13,09754 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...