Magnesium sulfate 2.25% (w/v) in aqueous solution

Supplier: Ricca Chemical

R4530000-500A R4530000-1A
RC453016EA 66.36 CAD
RC453016 RC453032
Magnesium sulfate 2.25% (w/v) in aqueous solution
Magnesium sulphate

APHA for BOD.

Formula: MgSO₄
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 2.250 ± 0.003 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR