Synthware Four-Neck Round-Bottom Flasks, Angled, Kemtech America
Supplier: Kemtech America
F221125 F221150 F221100 F221250 F224250 F221500 F224500 F222500 F224000
60002-864EA 192.36 CAD
60002-864 60002-868 60002-872 60002-876 60002-880 60002-882 60002-886 60002-888 60002-896
Synthware Four-Neck Round-Bottom Flasks, Angled, Kemtech America
Flasks Round Bottom Flasks
Heavy wall design.
  • Three angled side necks
  • Ground glass joints
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes available
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.