D-(-)-Mannitol, powder, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

MACR6209-04 MACR6209-05
CAMK620904CS 8943.5 CAD
CAMK620904 CAMK620905
D-(-)-Mannitol, powder, AR® ACS, Macron Fine Chemicals™
D(-)-Mannitol

Powder.

Formula: HOH₂C(CH(OH))₄CH₂OH
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.01 %
ACS - Loss on Drying at 105°C ≤0.05 %
ACS - Reducing Sugars Passes Test
ACS - Residue after Ignition ≤0.01 %
ACS - Titrable Acid (meq/g) ≤0.0008
ACS - Specific Rotation [Å]²⁵^D (+) 23.3 - 24.3 Degree
ACS - Melting Point 166 - 168 °C
Solubility (2 in 20) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR