Synthware Flat-Bottom Flasks, Kemtech America
Supplier: Kemtech America
F122450 F124100 F124125 F124250 F124300 F124500 F124000 F128000
60002-228EA 57.71 CAD
60002-228 60002-230 60002-232 60002-234 60002-238 60002-242 60002-246 60002-248
Synthware Flat-Bottom Flasks, Kemtech America
Flasks Round Bottom Flasks
Single-neck, flat-bottom flasks with an outer joint.
  • Single neck
  • Ground glass joint
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
  • Flat bottom
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.