PYREX® Round-Bottom Boiling Flasks, Long Neck, Corning
Supplier: Corning
4280-2L 4280-500 4280-5L 4280-250 4280-1L
89091-070CS 3326.53 CAD
89091-070 89091-292 89091-416 89091-470 29121-109 29121-163
PYREX® Round-Bottom Boiling Flasks, Long Neck, Corning
Flasks Round Bottom Flasks
Flasks have tooled mouths for uniform stopper fit and carefully controlled wall thicknesses to maintain proper balance between thermal expansion and mechanical strength for optimal shock resistance
They are extremely chemically durable for contaminant-free results and long life under rigid lab conditions. Working temperature: 230°C (446°F).