Petroleum spirit, 60…90 °C, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

PX0427-30 PX0427-5 PX0427-3
CAPX0427-30EA 2418 CAD
CAPX0427-30 CAPX0427-5 CAPX0427-3
Petroleum spirit, 60…90 °C, Supelco®
Petroleum spirit

Reagent

Boiling Pt: 60…90 °C
Density: 0.73 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Acid To pass test
Appearance To pass test
Boiling range 60-90°C
Color (APHA) 10 max
Heavy oils and fats To pass test
Odor To pass test
Residue after evaporation 0.001% max
Sulfur compounds (Doctor test) Negative


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR