Petroleum spirit, 35…60 °C, GR ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: Petroleum etherGasolinePetrolBenzine

PX0425-5 PX0425T-4 PX0425-3 PX0425-20 PX0425-2 PX0425-20 PX0425T-4 PX0425P-4 PX0425P-4 PX0425-20 PX0425-5 PX0425-3 PX0425-6 PX0425-2
CAPX0425P-4CS 2159.61 CAD
CAPX0425P-4 CAPX0425T-4 EMD-PX0425P-4 EM-PX0425-20 CAPX0425-2 CAPX0425-3 CAPX0425-5 CAPX0425-6 CAPX0425-20 EM-PX0425T-4 EM-PX0425-3 EM-PX0425-2 TXPX0425-20BSH EM-PX0425-5
Petroleum spirit, 35…60 °C, GR ACS, Supelco®
Petroleum spirit

Meets ACS Specifications

Boiling Pt: 30…50 °C (1013 hPa)
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3,I

Order Now

Specification Test Results

Acidity To pass test
Boiling range 35-60°C
Color (APHA) 10 max
Form Clear liquid
Residue after evaporation 0.001% max
Water (H2O) 0.01% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR