Magnesium chloride hexahydrate 99.0-102.0%, crystals, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Magnesium dichloride

MACR5958-26 MACR5958-06 5958-04 MACR5958-04 5958-06 MACR5958-26
CAMK595804CS 2920.99 CAD
MK595806 CAMK595804 CAMK595806 MK5958-04 MK5958-26 CAMK5958-26
Magnesium chloride hexahydrate 99.0-102.0%, crystals, AR® ACS, Macron Fine Chemicals™
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
MDL Number: MFCD00149781
CAS Number: 7791-18-6
Merck Index: 13,05684

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (MgCl₂·6H₂O) 99.0 - 102.0 %
ACS - Ammonium (NH₄) ≤0.001 %
ACS - Barium (Ba) ≤0.005 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Manganese (Mn) ≤5.000 ppm
ACS - Nitrate (NO₃) ≤0.001 %
ACS - pH of 5% Solution 4.5 - 7.0
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Potassium (K) ≤0.005 %
ACS - Sodium (Na) ≤0.005 %
ACS - Strontium (Sr) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.002 %
The net weight of the product in each container is within +/- 2% of the netweight stated on the label.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR