n-Pentane ≥98% (by GC), Supelco®

Supplier: MilliporeSigma
Danger

PX0170-3 PX0170-30 PX0170-1 PX0170-3
CAPX0170-3EA 1975 CAD
CAPX0170-3 CAPX0170-30 CAPX0170-1 EM-PX0170-3
n-Pentane ≥98% (by GC), Supelco®
n-Pentane

Store in a cool place.

Formula: H₃C(CH₂)₃CH₃
Boiling Pt: 35…36 °C (lit.)
Melting Pt: −130 °C (lit.)
Density: ~0.63 g/cm3 (20 °C), 0.626 g/ml (25°C) (lit.)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 98% min
Color (APHA) 30 max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 0.001% max
Water (H2O) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR