Acrylic Dome Mirrors, FRED SILVER & CO., INC.
Supplier: Fred Silver & Co.
DOME-X-18 DOME-X-26 DOME-X-32 DOME-X-36 H-DOME-18 H-DOME-26 H-DOME-36 Q-DOME-18 Q-DOME-26 Q-DOME-32
28801-060EA 409.79 CAD
28801-060 28801-062 28801-064 28801-066 28801-070 28801-072 28801-076 28801-080 28801-082 28801-084
Acrylic Dome Mirrors, FRED SILVER & CO., INC.
Mirrors
Safety/security mirrors are constructed of lightweight, 0.32cm (1/8") thick acrylic
Full dome mirrors provide a full 360° view from any angle. Ideal for four-way intersections. Supplied with hanging wires for suspension from the ceiling. Can also be attached directly to the ceiling.
Half dome (180°) mirrors are recommended for three-way (“T”) intersections. Can be wall or ceiling mounted.
Quarter dome (90°) mirrors are recommended for two-way (“L”) intersections. Screw holes are provided for corner/ceiling mounting and spring for corner/wall mounting.
Durable mirrors areMore Product Information