Mineral oil, white (petroleum), Supelco®

Supplier: MilliporeSigma
Warning

PX0045-3 PX0045-5
CA-PX0045-3EA 222 CAD
CA-PX0045-3 CAPX0045-5
Mineral oil, white (petroleum), Supelco®
White mineral oil (petroleum)
Boiling Pt: 300 °C (1013 hPa)
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
EINECS: 232-455-8

Order Now

Specification Test Results

Color (APHA) 20 max
Description To pass test
Form Liquid
Infrared Spectrum Conforms to standard
Limit of polynuclear compounds To pass test
Neutrality To pass test
Readily carbonizable substances To pass test
Solid paraffin To pass test
Solubility To pass test
Specific Gravity at 25/25°C 0.864-0.881


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR