Packer Bottles, Vacuum and Ionized, Amber, Wide Mouth, Qorpak
Supplier: Qorpak
GLC-02080 GLC-07007 GLC-08685 GLC-02107 GLC-02204 GLC-02126 GLC-02149 GLC-02083 GLC-02098 GLC-02192 GLC-02120 GLC-02137 GLC-02164 GLC-11310 GLC-02116 GLC-07100 GLC-07101 GLC-02189 GLC-02133 264628 264784 260311 260310 260327 260317 260304 260322 260324 260313 242090 242091 242095 242096 GLC-02073 GLC-07286
11311-178CS 2055.5 CAD
11311-178 11311-182 11311-382 11311-594 36319-728 36319-729 36319-730 36319-731 36319-732 36319-733 36319-774 36319-775 36319-776 36319-777 36319-778 36319-779 89092-718 93000-936 93000-940 93000-942 93000-950 93000-952 CA10736-442 10818-464 10818-466 10818-470 10818-472 10818-474 10818-476 10818-478 10818-480 10818-482 10818-484 76318-458 76318-460 76318-462 76318-464 76318-466 76487-302 76487-304 76487-306
Packer Bottles, Vacuum and Ionized, Amber, Wide Mouth, Qorpak
Bottles Glass Bottles
The vacuum and ionized cleaning process removes contaminants such as loose dirt, carton lint, aerosols, and fine glass particles.
  • Saves time, no need to waste valuable time rinsing and drying bottles
  • Seals cleanliness into each individual bottle
Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Then the air passes through an absorbent filter to remove gases. A vacuum then draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. The air inside the bottle will not interfere with your product in any way.