Toluene-α-sulfonyl fluoride ≥99.0%, OmniPur®, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulfonyl fluoride, α-Toluenesulphonyl fluoride, Phenylmethanesulfonyl fluoride, Phenylmethylsulfonyl fluoride

7110-5GM 7130-250GM
CA-EM7110EA 206.44 CAD
CA-EM7110 CAEM7130
Toluene-α-sulfonyl fluoride ≥99.0%, OmniPur®, Sigma-Aldrich®
Toluene-α-sulphonyl fluoride

White powder. Insoluble in water.

Formula: C₇H₇FO₂S
MW: 174.2 g/mol
Boiling Pt: 80…85 °C (0.3 mmHg)
Melting Pt: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR