Magnesium sulfate heptahydrate 99,5, GenAR® USP Parenteral Use, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

M.O.C. Category R

MACR4200-07 MACR4200-20
CAMK420007EA 883.9 CAD
CAMK420007 CAJT4200-16 CAMK420020
Magnesium sulfate heptahydrate 99,5, GenAR® USP Parenteral Use, Macron Fine Chemicals™
Magnesium sulphate heptahydrate

Crystals.

Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (as MgSO₄) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - pH (1 in 20) 5.0 - 8.8
USP - Loss on Ignition 40.0 - 52.0 %
USP - Chloride (Cl) ≤ 0.014 %
USP - Selenium (Se) ≤ 0.003 %
USP - Iron (Fe)(µg/g) ≤ 0.5
Solubility (5 in 50) Passes Test
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR