2-Methylbutane ≥95%, OR, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: iso-Pentane

Management of Change (MOC) category = N

MACR1967-59 1967-59
CAMK196759EA 207.33 CAD
CAMK196759 MK196759
2-Methylbutane ≥95%, OR, Macron Fine Chemicals™
2-Methylbutane

Macron Fine Chemicals™ OR: organic reagents that are suitable for research applications

Formula: CH₃CH₂CH(CH₃)₂
MW: 72.15 g/mol
Boiling Pt: 28 °C (1013 hPa)
Melting Pt: –160 °C
Density: 0.62 g/cm³ (20 °C)
Flash Pt: –51 °C
MDL Number: MFCD00009338
CAS Number: 78-78-4
UN: 1265
ADR: 3,I
https://ca.vwr.com/stibo/web/std.lang.all/04/68/740468.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay ((CH₃)₂CHCH₂CH₃) ≥95 %
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR