Readycult® Sample Bottles and Coliform Tests, MilliporeSigma
Supplier: MilliporeSigma
Readycult®
1.120SB.0101 1.120SB.0102
CA1120SB.0101EA 472.68 CAD
CA1120SB.0101 CA1120SB.0102
Readycult® Sample Bottles and Coliform Tests, MilliporeSigma
Bottles Water Sampling Bottles
EPA-approved method for detecting the presence or absence of total coliforms and E.coli.
  • Presence of total coliforms is indicated by a blue-green color of the broth
  • Presence of E. coli is indicated by a blue fluorescence under UV light
Dehydrated and granulated culture media in an irradiated snap pack format is added to the water sample and incubated for 24 hours at 34.5 to 37 °C (94.1 to 98.6 °F).
Sample bottles are sterile and are available with or without sodium thiosulfate dechlorinating agent.