2-Propanol 99,5 ACS zA, General Laboratory Use

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

LP323-19 LP323-4 LP323-1P LP323-204 LP323-4P LP323-5G
CABJLP323D19EA 1443.24 CAD
CABJLP323D19 CABJLP323-4 BJLP323-1P BJLP323-204 BJLP323-4P BJLP323-5G
2-Propanol 99,5 ACS zA, General Laboratory Use
2-Propanol

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.785 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis 99.5 % min.
Carbonyl Compounds 0.002 % max.
Color (APHA) 10 max.
Residue after Evaporation 0.001 % max.
Solubility in Water Passes test
Titrable Acid or Base 0.1 µeq/g max.
Water by Karl Fishcer Titration 0.2 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR