2-Methylbutane ≥99.0%, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: iso-Pentane

Management of Change (MOC) category = N

BAKRQ223-07 Q223-07 BAKRQ223-08 Q223-08
CAJTQ223-8CS 2479.35 CAD
JTQ223-8 JTQ223-7 CAJTQ223-8 CAJTQ223-7
2-Methylbutane ≥99.0%, J.T.Baker®
2-Methylbutane
Formula: CH₃CH₂CH(CH₃)₂
MW: 72.15 g/mol
Boiling Pt: 28 °C (1013 hPa)
Melting Pt: –160 °C
Density: 0.62 g/cm³ (20 °C)
Flash Pt: –51 °C
MDL Number: MFCD00009338
CAS Number: 78-78-4
UN: 1265
ADR: 3,I
https://ca.vwr.com/stibo/web/std.lang.all/04/68/740468.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Store at or below 68°F (20°C).
Assay ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR