Pyridine 99,8, BioSyn® Biochemistry, water_low_applications, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Azabenzene, Azine

BB333-4 BC333-200 BB333-2.5L
CABJBB333-4CS 6499.74 CAD
CABJBB333-4 BJBC333-200 BJBN333-20 CABJBB333-2.5L
Pyridine 99,8, BioSyn® Biochemistry, water_low_applications, Burdick & Jackson™
Pyridine

BioSyn« products are ideal for DNA, RNA and oligonucleotide synthesis. All solvents and regents are filtered (0.2-micron) and tested for oxidation reage.


  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity

Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115
Melting Pt: -44
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3 II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Amines by Ninhydrin Test 10 ppm max.
Free Acid Passes test
GC Analysis 99.8 % min.
Refractive Index (20ºC) 1.5085 - 1.5109
Residue 1 mg/L max.
Water by Karl Fischer Titration 50 ppm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR