2-Propanol ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

AH323-4PC AH323-4
CABJAH323-4PCCS 1082.6 CAD
CABJAH323-4PC CABJAH323-4
2-Propanol ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
2-Propanol

Meets USP Reagent Specifications


  • ACS certified
  • Lot-to-lot consistency
  • HPLC grade controlled for UV absorbance, GC assay, water content and non-volatile residue

Formula: (CH₃)₂CHOH
Boiling Pt: 12.0 %(V)
Melting Pt: –88 °C
Density: 0.785 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Carbonyl Compounds 0.002 % max.
Water by Karl Fischer Titration 0.06 % max.
Color (APHA) 10 max.
GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.3766 - 1.3778
Residue 3 mg/L max.
Titrable Acid or Base 0.1 µeq/g max.
UV Absorbance @ 225 nm 0.16 AU max.
UV Absorbance @ 250 nm 0.02 AU max.
UV Cutoff 205 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR