Maleic acid ≥99% (by acidimetry), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKRP460-07 BAKRP460-08
CAJTP460-7EA 289.96 CAD
CAJTP460-7 CAJTP460-8
Maleic acid ≥99% (by acidimetry), J.T.Baker®
Maleic acid
Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 130…131 °C
Density: 1.59 g/cm³ (20 °C)
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
EINECS: 203-742-5
UN: 3261
ADR: 8,III
Merck Index: 13,05726
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a1ea1f5bca5d5cf7fd40530f642f47db88908a&vid=1ac7f8cee2cd0433dbfb6e9271d41fe6ecc382c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₄H₄O₄) (by acidimetry) ≥99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR