Methanol ≥99.9%, BAKER ANALYZED® LC-MS Reagent for LC-MS, for HPLC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Management of Change (MOC) category = N

BAKR9830-03 9830-02 9830-03
CAJT9830-2CS 7775.5 CAD
CAJT9830-2 CAJT9830-3 JT9830-2 JT9830-3
Methanol ≥99.9%, BAKER ANALYZED® LC-MS Reagent for LC-MS, for HPLC, J.T.Baker®
Methanol

Pure, trusted chromatography solvents providing high selectivity, reproducibility and accuracy of results that come in a variety of grades such as HPLC, LCMS, ULTRA LCMS and PHOTREX.

Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃OH) (by GC, corrected for water) ≥99.9 %
Appearance Passes Test
Gradient Elution Test (a.u.) - 254 nm ≤0.01
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Water (H₂O) ≤500 ppm
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤50 ppb
Trace Impurities - Aluminum (Al) ≤50.0 ppb
Trace Impurities - * Calcium (Ca) ≤50.0 ppb
Trace Impurities - Iron (Fe) ≤50.0 ppb
Trace Impurities - Lithium (Li) ≤30.0 ppb
Trace Impurities - Magnesium (Mg) ≤50.0 ppb
Trace Impurities - Nickel (Ni) ≤30.0 ppb
Trace Impurities - Potassium (K) ≤50 ppb
Trace Impurities - * Sodium (Na) ≤50.0 ppb
* May change over time due to extraction from glass container.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR