2-Propanol ≥99.9%, B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

323-1L 323-4 NP323-20 NS323-20
CABJ323-1LCS 537.52 CAD
CABJ323-1L CABJ323-4 CABJNP323-20 BJNS323-20
2-Propanol ≥99.9%, B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™
2-Propanol

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.785 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC 2 µg/L max.
Water by Karl Fischer Titration 0.06 % max.
GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.3766 - 1.3778
Residue 1 mg/L max.
UV Absorbance @ 225 nm 0.16 AU max.
UV Absorbance @ 250 nm 0.02 AU max.
UV Absorbance @ 300 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 205 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR