1-Propanol ≥99.5%, B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
322-1L
322-4
CABJ322-1LCS
1027.36
CAD
CABJ322-1L
CABJ322-4
1-Propanol ≥99.5%, B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™
1-Propanol
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Formula:
H₃CCH₂CH₂OH Boiling Pt: 97.2 °C Melting Pt: –126.2 °C Density: 0.8053 g/cm³ (20 °C) Flash Pt: 13.7 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00002941 CAS Number: 71-23-8 EINECS: 200-746-9 UN: 1274 ADR: 3,III Merck Index: 13,07934 |
|
Specification Test Results
GC Analysis | 99.5 % min. |
Refractive Index (20ºC) | 1.3841 - 1.3857 |
Residue | 3 mg/L max. |
UV Absorbance @ 225 nm | 0.5 AU max. |
UV Absorbance @ 250 nm | 0.05 AU max. |
UV Absorbance @ 300 nm | 0.005 AU max. |
UV Absorbance @ 400 nm | 0.005 AU max. |
UV Cutoff | 210 nm max. |
Water by Karl Fischer Titration | 0.05 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...