4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
BAKR9322-01
BAKR9322-03
CAJT9322-1CS
2537.76
CAD
CAJT9322-1
CAJT9322-3
4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), BAKER ANALYZED® ACS
4-Methyl-2-pentanone
Lot analysis on label.
Formula:
(CH₃)₂CHCH₂COCH₃ Boiling Pt: 117.4 °C (1013 hPa) Melting Pt: –84 °C Density: 0.801 g/cm³ (20 °C) Flash Pt: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008938 CAS Number: 108-10-1 EINECS: 203-550-1 UN: 1245 ADR: 3,II Merck Index: 13,05227 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water) | ≥98.5 % |
Color (APHA) | ≤15 |
Residue after Evaporation | ≤0.0050 % |
Titrable Acid (meq/g) | ≤0.002 |
Water (H₂O)(by Karl Fischer titrn) | ≤0.1 % |
Product Information (not specifications): | |
Appearance (Clear, colorless liquid) | |
Boiling Point (typical)................. 115.5 °C. | |
Density (g/mL) at 25 °C (typical)........ 0.796 | |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...