Ethanolamine ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
BAKR9314-01
BAKR9314-03
CAJT9314-1CS
5266
CAD
CAJT9314-1
CAJT9314-3
Ethanolamine ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Ethanolamine
Lot analysis on label.
Formula:
NH₂CH₂CH₂OH Boiling Pt: 171 °C (1013 hPa) Melting Pt: 10.5 °C Density: 1.0164 g/cm³ (20 °C) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008183 CAS Number: 141-43-5 EINECS: 205-483-3 UN: 2491 ADR: 8,III Merck Index: 13,03762 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (HOCH₂CH₂NH₂) (by GC, corrected for water) | ≥ 99.0 % |
Color (APHA) | ≤ 15 |
Water (H₂O) | ≤ 0.30 % |
Heavy Metals (by ICP-OES) | ≤ 5 ppm |
Trace Impurities - Iron (Fe) | ≤ 5 ppm |
Meets Reagent Specifications for testing USP/NF monographs Product | |
Information (not specifications):Density (g/mL) @ 25°C (typical)=1.012 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...