N-Methyl-2-pyrrolidone (NMP) ≥99.5%, BAKER BIO-ANALYZED® for biotechnology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

Management of Change (MOC) category = N

BAKR9261-09 BAKR9261-13
CAJT9261-9EA 11837.22 CAD
CAJT9261-9 CAJT9261-13
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, BAKER BIO-ANALYZED® for biotechnology, J.T.Baker®
N-Methyl-2-pyrrolidone (NMP)

For Biotechnology applications. UV Cut-off 285nm.


  • UV Cut-off 285nm

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 (open cup)
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₅H₉NO) ≥99.5 %
Color (APHA) ≤20
Free Amines (as CH₃NH₂) ≤0.010 %
Heavy Metals (as Pb) ≤0.10 ppm
Residue after Ignition ≤10 ppm
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm ≤0.010
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.03
Ultraviolet Absorbance (1.00-cm cell vs. water) - 325 nm ≤0.100
Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm ≤0.5
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤285
Water (H₂O)(by coulometry) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR